Learn More
AdipoGen Pseurotin D
SDP
Catalog No. 502561933
Encompass_Preferred
Chemical. CAS 77409-68-8. Formula C22H25NO8. MW 431.4. Isolated from Aspergillus fumigatus. Isomeric to pseurotin A Prod. No. BVT-0003. Anticancer and antiparasitic compound. Apomorphine antagonist. Neuroleptic agent. Inhibitor of IgE production.
Catalog No. 50-256-1933 Supplier AdipoGen Supplier No. BVT0426M001
Chemical. CAS 77409-68-8. Formula C22H25NO8. MW 431.4. Isolated from Aspergillus fumigatus. Isomeric to pseurotin A Prod. No. BVT-0003. Anticancer and antiparasitic compound. Apomorphine antagonist. Neuroleptic agent. Inhibitor of IgE production.