Learn More
FUJIFILM Biosciences PSFM-J1 Medium Wako Liquid
Catalog No. 5000450408
Encompass_Preferred
This product is for culturing insect cells specifically Sf9 cells and H5 cells As a serum-free medium it eliminates concerns about variations in biological activity due to serum lots or viral contamination
Catalog No. 50-004-50408 Supplier FUJIFILM Biosciences Supplier No. 16025851
This product is for culturing insect cells specifically Sf9 cells and H5 cells As a serum-free medium it eliminates concerns about variations in biological activity due to serum lots or viral contamination