Learn More
Aviva Systems Biology Corp PSG1 Antibody - N-terminal region (ARP42325_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42325_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PSG1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PSG1. Alternate names: B1G1, CD66f, DHFRP2, FLJ90598, FLJ90654, PBG1, PSBG1, PSGGA, PSGIIA, SP1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-58184