Learn More
Aviva Systems Biology Corp PSG3 Antibody - N-terminal region (ARP42577_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42577_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PSG3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PSG3. Alternate names: ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-58273