Learn More
Aviva Systems Biology Corp PSG6 Antibody - N-terminal region (ARP41880_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41880_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSG6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PSG6. Alternate names: PSG10, PSBG-6, PSBG-10, PSBG-12ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58033