Learn More
Medchemexpress LLC PSG9 Human HEK293 10ug
SDP
Catalog No. 5000196883
Encompass_Preferred
Recombinant human PSG9 protein expressed in HEK293 mammalian cells and supplied as a C-terminal 6xHis-tagged, glycosylated solution formulated in 20 mM PB, 150 mM NaCl, pH 7.2. The preparation is >95% pure by reducing SDS-PAGE, has endotoxin <1 EU/μg, and is intended for research applications requiring a mammalian-expressed, post-translationally modified protein. Store at -80°C; shipped on dry ice.
  • Produced in HEK293 mammalian cells for authentic glycosylation.
  • C-terminal 6xHis tag to enable affinity purification and detection.
  • High purity (>95%) suitable for biochemical assays.
  • Low endotoxin (<1 EU/μg) for sensitive cell-based experiments.
  • Supplied in a filtered, ready-to-use buffered solution.
  • Shipped on dry ice and stable when stored at -80°C.
Catalog No. 50-001-96883 Supplier Medchemexpress LLC Supplier No. HYP7098410UG
Recombinant human PSG9 protein expressed in HEK293 mammalian cells and supplied as a C-terminal 6xHis-tagged, glycosylated solution formulated in 20 mM PB, 150 mM NaCl, pH 7.2. The preparation is >95% pure by reducing SDS-PAGE, has endotoxin <1 EU/μg, and is intended for research applications requiring a mammalian-expressed, post-translationally modified protein. Store at -80°C; shipped on dry ice.
  • Produced in HEK293 mammalian cells for authentic glycosylation.
  • C-terminal 6xHis tag to enable affinity purification and detection.
  • High purity (>95%) suitable for biochemical assays.
  • Low endotoxin (<1 EU/μg) for sensitive cell-based experiments.
  • Supplied in a filtered, ready-to-use buffered solution.
  • Shipped on dry ice and stable when stored at -80°C.