Learn More
ACROBiosystems ACROBiosystems Human PSGL-1 / CD162 Protein, His Tag
Catalog No. 501956427
Encompass_Preferred
Recombinant Protein;1MG;Human PSGL-1, His Tag (PS1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Cys 320 (Accession # Q14242-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.8 kDa.
Catalog No. 50-195-6427 Supplier ACROBiosystems Supplier No. PS1H52H91MG
Recombinant Protein;1MG;Human PSGL-1, His Tag (PS1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Cys 320 (Accession # Q14242-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.8 kDa.