Learn More
Sino Biological PSGL-1 / CD162 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 50770T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse PSGL-1 / CD162 (Catalog#50770-MCCH; Q62170; Met1-Cys307). PSGL-1 / CD162 specific IgG was purified by Mouse PSGL-1 / CD162 affinity chromatography.
Catalog No. 50-224-8405