Learn More
New England Biolabs, Inc. PshAI – 1000 units
SDP
Catalog No. 50812308
Encompass
A restriction endonuclease that recognizes the sequence GACNN_^NNGTC.
Catalog No. 50-812-308 Supplier New England Biolabs, Inc. Supplier No. R0593S
A restriction endonuclease that recognizes the sequence GACNN_^NNGTC.