Learn More
Agilent Technologies PSHUTTLE-CMV LACZ VECTOR
Catalog No. 50125390
Encompass
PShuttle-CMV lacZ Vector is a control vector produced by insertion of the lacZ gene in the multiple cloning site of the pShuttle-CMV. This construct is provided as a control for the production of recombinant adenovirus
Catalog No. 50-125-390 Supplier Agilent Technologies Supplier No. 240008
PShuttle-CMV lacZ Vector is a control vector produced by insertion of the lacZ gene in the multiple cloning site of the pShuttle-CMV. This construct is provided as a control for the production of recombinant adenovirus