Learn More
Agilent Technologies PSHUTTLE VECTOR
Catalog No. 50125388
Encompass
pShuttle Vector is a containing only a multiple cloning site in which an entire expression cassette can be inserted.THis vector contains the kanamycin resistance gene for the selection of recombinants
Catalog No. 50-125-388 Supplier Agilent Technologies Supplier No. 240006
pShuttle Vector is a containing only a multiple cloning site in which an entire expression cassette can be inserted.THis vector contains the kanamycin resistance gene for the selection of recombinants