Learn More
Medchemexpress LLC HY-10165 2mg Medchemexpress, PSI-6130 CAS:817204-33-4 Purity:>98%
SDP
Catalog No. 501932190
Encompass_Preferred
Medchemexpress, HY-10165 2mg PSI-6130 CAS:817204-33-4 PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase , and inhibits HCV replication with a mean IC 50 of 0.6 μM.  Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2190 Supplier Medchemexpress LLC Supplier No. HY101652MG
Medchemexpress, HY-10165 2mg PSI-6130 CAS:817204-33-4 PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase , and inhibits HCV replication with a mean IC 50 of 0.6 μM.  Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.