Learn More
Medchemexpress LLC HY-15236S 1mg Medchemexpress, PSI-6206 13CD3 CAS:1256490-42-2 Purity:>98%
SDP
Catalog No. 501932727
Encompass_Preferred
Medchemexpress, HY-15236S 1mg PSI-6206 13CD3 CAS:1256490-42-2 PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC 90 of >100 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2727 Supplier Medchemexpress LLC Supplier No. HY15236S1MG
Medchemexpress, HY-15236S 1mg PSI-6206 13CD3 CAS:1256490-42-2 PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC 90 of >100 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.