Learn More
Selleck Chemical LLC PSI-6206 (GS-331007) S5307-25mg
SDP
Catalog No. 503109552
Encompass_Preferred
PSI-6206 (RO-2433 GS-331007) is a potent inhibitor of the hepatitis C virus RNA-dependent RNA polymerase targeting NS5B polymerase
Catalog No. 50-310-9552 Supplier Selleck Chemical LLC Supplier No. S530725MG
PSI-6206 (RO-2433 GS-331007) is a potent inhibitor of the hepatitis C virus RNA-dependent RNA polymerase targeting NS5B polymerase