Learn More
Medchemexpress LLC HY-15745A 1mg Medchemexpress, PSI-7409 tetrasodium CAS:1621884-22-7 Purity:>98%
SDP
Catalog No. 501969955
Encompass_Preferred
Medchemexpress, HY-15745A 1mg PSI-7409 tetrasodium CAS:1621884-22-7 PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9955 Supplier Medchemexpress LLC Supplier No. HY15745A1MG
Medchemexpress, HY-15745A 1mg PSI-7409 tetrasodium CAS:1621884-22-7 PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.