Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PSI-7976 | 1190308-01-0 | 98.2% | 529.45 g/mol | C22H29FN3O9P | 10 MG
SDP

Catalog No. 5000286175
Encompass_Preferred

PSI-7976 is the stereoisomer of a nucleotide analogue hepatitis C virus (HCV) RNA replication inhibitor supplied for research use. It is listed in product and database records with CAS 1190308-01-0, chemical formula C22H29FN3O9P, molecular weight 529.45 g/mol, and reported purity of about 98.2%. The compound is intended for in vitro assays and assay development and is not for human therapeutic use.

  • Isomer of a nucleotide analogue HCV RNA replication inhibitor
  • Intended for in vitro research and assay development
  • Reported purity approximately 98.2%
  • Molecular weight 529.45 g/mol
  • Chemical formula C22H29FN3O9P
  • CAS number 1190308-01-0

Catalog No. 50-002-86175 Supplier Medchemexpress LLC Supplier No. HY15005A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSI-7976 is the stereoisomer of a nucleotide analogue hepatitis C virus (HCV) RNA replication inhibitor supplied for research use. It is listed in product and database records with CAS 1190308-01-0, chemical formula C22H29FN3O9P, molecular weight 529.45 g/mol, and reported purity of about 98.2%. The compound is intended for in vitro assays and assay development and is not for human therapeutic use.

  • Isomer of a nucleotide analogue HCV RNA replication inhibitor
  • Intended for in vitro research and assay development
  • Reported purity approximately 98.2%
  • Molecular weight 529.45 g/mol
  • Chemical formula C22H29FN3O9P
  • CAS number 1190308-01-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.