Learn More
Medchemexpress LLC HY-15005A 1mg Medchemexpress, PSI-7976 CAS:1190308-01-0 Purity:>98%
SDP
Catalog No. 501932718
Encompass_Preferred
Medchemexpress, HY-15005A 1mg PSI-7976 CAS:1190308-01-0 PSI-7976 is the isomer of PSI-7977. PSI-7977 is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus ( HCV ) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2718 Supplier Medchemexpress LLC Supplier No. HY15005A1MG
Medchemexpress, HY-15005A 1mg PSI-7976 CAS:1190308-01-0 PSI-7976 is the isomer of PSI-7977. PSI-7977 is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus ( HCV ) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.