Learn More
Cayman Chemical PSI-7977-13C-d3 1mg
SDP
Catalog No. 502933963
Encompass_Preferred
An internal standard for the quantification of PSI-7977 by GC- or LC-MS
Catalog No. 50-293-3963 Supplier Cayman Chemical Supplier No. 250451 MG
Add to Cart
Add to Cart
An internal standard for the quantification of PSI-7977 by GC- or LC-MS