Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Psi Reagent

Catalog No. ALD0060225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD0060225G 25 g
ALD00602100 100 g
ALD006025G 5 g
ALD00602 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ALD0060225G Supplier Sigma Aldrich Supplier No. ALD0060225G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-Psi Reagent

Specifications

CAS 2245335-71-9
Melting Point 102°C to 107°C
Molecular Formula C16H16F5OPS3
Linear Formula C16H16F5OPS3
Quantity 25 g
Synonym (+)-Psi; (2R,3aR,6S,7aR)-3a-Methyl-2-((perfluorophenyl)thio)-6-(prop-1-en-2-yl)hexahydrobenzo[d][1,3,2]oxathiaphosphole 2-sulfide; Phosphorus-Sulfur Incorporation Reagent
Molecular Weight (g/mol) 446.46 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.