Learn More
Medchemexpress LLC HY-119819 1mg Medchemexpress, Psicofuranine CAS:1874-54-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1198191MG
Encompass_Preferred
Medchemexpress, HY-119819 1mg Psicofuranine CAS:1874-54-0 Psicofuramine a nucleoside antibiotic and has the inhibition of xanthosine 5'-phosphate aminase. Psicofuranine also specifically inhibits GMP synthase, and interrupts parasite growth. Psicofuranine exhibits a dose-dependent inhibition of P. falciparum growth. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9548