Learn More
System Biosciences LLC PSIH1-H1-SILUC-COP-GFP 10UGpSIH1-H1-siLuc-copGFP positive transduction control, plasmid = 10ug
SDP
Catalog No. NC3306486
Encompass_Preferred
Test the sensitivity of your target cells to HIV-based lentiviral transduction and get quantitative measurements of transduction efficiencies using copGFP. Some cell lines are transduced less efficiently than others. To help with lentiviral transduction optimization and troubleshooting, SBI offers a series of positive transduction controls that come as pre-packaged, ready-to-transduce lentiviral particles as well as ready-to-transfect and package lentivector plasmids. The pSIH1-H1-siLuc-copGFP Positive Transduction Control enables quantitation of HIV-based lentiviral transduction efficiency using the bright copGFP protein.
Catalog No. NC3306486 Supplier System Biosciences LLC Supplier No. LV601PB1
Test the sensitivity of your target cells to HIV-based lentiviral transduction and get quantitative measurements of transduction efficiencies using copGFP. Some cell lines are transduced less efficiently than others. To help with lentiviral transduction optimization and troubleshooting, SBI offers a series of positive transduction controls that come as pre-packaged, ready-to-transduce lentiviral particles as well as ready-to-transfect and package lentivector plasmids. The pSIH1-H1-siLuc-copGFP Positive Transduction Control enables quantitation of HIV-based lentiviral transduction efficiency using the bright copGFP protein.