Learn More
New England Biolabs, Inc. PsiI-v2 – 2000 units
SDP
Catalog No. NC1694786
Encompass
A restriction endonuclease that recognizes the sequence TTA_^TAA
Catalog No. NC1694786 Supplier New England Biolabs, Inc. Supplier No. R0744L
A restriction endonuclease that recognizes the sequence TTA_^TAA