Learn More
New England Biolabs, Inc. PsiI-v2 – 400 units
SDP
Catalog No. NC1704764
Encompass
A restriction endonuclease that recognizes the sequence TTA_^TAA
Catalog No. NC1704764 Supplier New England Biolabs, Inc. Supplier No. R0744S
A restriction endonuclease that recognizes the sequence TTA_^TAA