Learn More
Aviva Systems Biology Corp PSIP1 Antibody - N-terminal region (ARP34753_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34753_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PSIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PSIP1. Alternate names: DFS70, LEDGF, MGC74712, PAIP, PSIP2, p52, p75ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-55546