Learn More
Aviva Systems Biology Corp PSIP1 Antibody - middle region (ARP34644_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34644_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PSIP1. Alternate names: p52, p75, PAIP, DFS70, LEDGF, PSIP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-55496