Learn More
Medchemexpress LLC PSMA-1007 | 2226894-58-0 | C₅₄H₆₄F₃N₉O₁₈ | 5 MG
SDP
Catalog No. 5000372714
Encompass_Preferred
PSMA-1007 is a prostate-specific membrane antigen (PSMA) ligand used for research. It can be used as a PET tracer for prostate cancer imaging and for the synthesis/research of Radionuclide-Drug Conjugates (RDCs).
  • Prostate-specific membrane antigen (PSMA) ligand
  • PET tracer for prostate cancer imaging
  • Used in synthesis/research of radionuclide-drug conjugates (RDCs)
  • Shows high uptake in tumor sites in prostate cancer xenograft mouse models
Catalog No. 50-003-72714 Supplier Medchemexpress LLC Supplier No. HY1535505MG
Add to Cart
Add to Cart
PSMA-1007 is a prostate-specific membrane antigen (PSMA) ligand used for research. It can be used as a PET tracer for prostate cancer imaging and for the synthesis/research of Radionuclide-Drug Conjugates (RDCs).
  • Prostate-specific membrane antigen (PSMA) ligand
  • PET tracer for prostate cancer imaging
  • Used in synthesis/research of radionuclide-drug conjugates (RDCs)
  • Shows high uptake in tumor sites in prostate cancer xenograft mouse models