Learn More
Medchemexpress LLC PSMA-11 (HBED-CC-PSMA) | 1366302-52-4 | 99.7% | 946.99 | C44H62N6O17 | 10MG
SDP
Catalog No. 5000235083
Encompass_Preferred
PSMA-11 (HBED-CC-PSMA) is a PSMA-targeting ligand used as a precursor for radiolabeled PET tracers. Supplied as a white to off-white solid, it is intended for radiolabeling with gallium-68 or other radionuclides for preclinical and research PET imaging of PSMA-expressing prostate cancer. The compound's high purity and chelator-conjugated design support efficient tracer preparation and consistent labeling performance.
  • PSMA-targeting ligand for PET tracer development.
  • Contains HBED-CC chelator for efficient radiolabeling.
  • High purity (99.7%) suitable for radiolabeling applications.
  • Supplied as a solid for synthesis and formulation workflows.
  • Available in multiple pack sizes to suit research needs.
  • Used for preclinical imaging of PSMA-expressing tumors.
Catalog No. 50-002-35083 Supplier Medchemexpress LLC Supplier No. HY12539910MG
PSMA-11 (HBED-CC-PSMA) is a PSMA-targeting ligand used as a precursor for radiolabeled PET tracers. Supplied as a white to off-white solid, it is intended for radiolabeling with gallium-68 or other radionuclides for preclinical and research PET imaging of PSMA-expressing prostate cancer. The compound's high purity and chelator-conjugated design support efficient tracer preparation and consistent labeling performance.
  • PSMA-targeting ligand for PET tracer development.
  • Contains HBED-CC chelator for efficient radiolabeling.
  • High purity (99.7%) suitable for radiolabeling applications.
  • Supplied as a solid for synthesis and formulation workflows.
  • Available in multiple pack sizes to suit research needs.
  • Used for preclinical imaging of PSMA-expressing tumors.