Learn More
Medchemexpress LLC Psma-11 (HBED-CC-PSMA) | 1366302-52-4 | 99.7% | 946.99 g/mol | C44H62N6O17 | 5MG
SDP
Catalog No. 5000236033
Encompass_Preferred
PSMA-11 (HBED-CC-PSMA) is a small-molecule ligand that binds prostate-specific membrane antigen (PSMA). Supplied as a research reagent, it is used as a precursor for radiolabeling with gallium-68 to produce 68Ga-PSMA-11 for PET imaging of prostate cancer. The material is provided as a solid for laboratory use and should be stored protected from light at the recommended temperature.
Catalog No. 50-002-36033 Supplier Medchemexpress LLC Supplier No. HY1253995MG
PSMA-11 (HBED-CC-PSMA) is a small-molecule ligand that binds prostate-specific membrane antigen (PSMA). Supplied as a research reagent, it is used as a precursor for radiolabeling with gallium-68 to produce 68Ga-PSMA-11 for PET imaging of prostate cancer. The material is provided as a solid for laboratory use and should be stored protected from light at the recommended temperature.