Learn More
ACROBiosystems ACROBiosystems Cynomolgus PSMA / FOLH1 Protein, His Tag
Catalog No. 501956522
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus PSMA, His Tag (PSA-C5247) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # A0A2K5VNZ0-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.4 kDa.
Catalog No. 50-195-6522 Supplier ACROBiosystems Supplier No. PSAC52471MG
Recombinant Protein;1MG;Cynomolgus PSMA, His Tag (PSA-C5247) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # A0A2K5VNZ0-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.4 kDa.