Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human PSMA / FOLH1 Protein, His Tag
Catalog No. 501956701
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human PSMA, His Tag (PSA-HF244) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1). It is the FITC labeled form of Human PSMA, His Tag (PSA-H82Qb).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.4 KDa. The protein migrates as 85-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6701 Supplier ACROBiosystems Supplier No. PSAHF244200UG
Recombinant Protein;200UG;FITC-Labeled Human PSMA, His Tag (PSA-HF244) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1). It is the FITC labeled form of Human PSMA, His Tag (PSA-H82Qb).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.4 KDa. The protein migrates as 85-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.