Learn More
ACROBiosystems ACROBiosystems Human PSMA / FOLH1 Protein, His Tag
Supplier:  ACROBiosystems PSAH52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1 ).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.4 kDa. ;Immobilized Monoclonal Anti-Human PSMA Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind Human PSMA, His Tag (Cat. No. PSA-H52H3) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-195-6983