Prostate-specific membrane antigen (PSMA), also known as FOLH1, is a type II transmembrane zinc metallopeptidase belonging to the peptidase M28 family. This recombinant protein is expressed in human HEK293 cells and contains amino acids Lys 44 - Ala 750 with a polyhistidine tag at the N-terminus. It exhibits both folate hydrolase and N-acetylated-alpha-linked acidic dipeptidase (ALADase) activities. The protein is verified by mass spectrometry and SEC-MALS to ensure high quality and activity.
- Greater than 95% purity as determined by SDS-PAGE
- Calculated molecular weight of 81.4 kDa, migrating as 95-115 kDa under reducing conditions
- Verified substrate hydrolysis activity of >200 pmol/min/μg
- Endotoxin level less than 1.0 EU per μg
- Lyophilized from a 0.22 μm filtered solution in 25 mM MES, 500 mM NaCl, pH 6.5 with trehalose