Learn More
ACROBiosystems ACROBiosystems Biotinylated Human PSMA / FOLH1 Protein, His,Avitag™
Supplier:  ACROBiosystems PSAH82QB25UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human PSMA, His,Avitag (PSA-H82Qb) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1 ).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 83.2 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human PSMA Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human PSMA, His,Avitag (Cat. No. PSA-H82Qb) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-195-6803