Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PSMA I&S TFA | 98.2% | 1413.43 | 1 MG
SDP

Catalog No. 5000363116
Encompass_Preferred

PSMA I&S (TFA) is a precursor of the 99mTc-labeled PSMA-targeting ligand, appearing as a white to off-white solid.

  • Target PSMA
  • Inhibits immunology/inflammation pathway
  • Soluble in DMSO (100 mg/mL)
  • Soluble in various in vivo dissolution methods
  • Store at -20°C, protected from light and under nitrogen

Catalog No. 50-003-63116 Supplier Medchemexpress LLC Supplier No. HY148152A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSMA I&S (TFA) is a precursor of the 99mTc-labeled PSMA-targeting ligand, appearing as a white to off-white solid.

  • Target PSMA
  • Inhibits immunology/inflammation pathway
  • Soluble in DMSO (100 mg/mL)
  • Soluble in various in vivo dissolution methods
  • Store at -20°C, protected from light and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.