Learn More
Sino Biological Human PSMA / FOLH1 Protein (ECD,His Tag) 1mg
Supplier:  Sino Biological 15877H07H1
Encompass_Preferred
A DNA sequence encoding the human FOLH1 (NP_004467.1) (Lys44-Ala750) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-211-5438