Learn More
Aviva Systems Biology Corp PSMA1 Antibody - C-terminal region (ARP40417_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40417_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PSMA1. Alternate names: HC2, MGC14542, MGC14575, MGC14751, MGC1667, MGC21459, MGC22853, MGC23915, NU, PROS30ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57438