Learn More
Aviva Systems Biology Corp PSMA2 Antibody - N-terminal region (ARP56453_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56453_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PSMA2. Alternate names: HC3, MU, PMSA2, PSC2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61865