Learn More
Aviva Systems Biology Corp PSMA3 Antibody - middle region (ARP57743_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57743_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PSMA3. Alternate names: HC8, MGC12306, MGC32631, PSC3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62140