Learn More
Aviva Systems Biology Corp PSMA5 Antibody - N-terminal region (ARP56455_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56455_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PSMA5. Alternate names: MGC117302, MGC125802, MGC125803, MGC125804, PSC5, ZETAApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61866