Learn More
Aviva Systems Biology Corp PSMB9 Antibody - C-terminal region (ARP46081_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46081_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMB9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PSMB9. Alternate names: LMP2, MGC70470, RING12, PSMB6i, beta1iApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59215