Learn More
Aviva Systems Biology Corp Psmc1 Antibody - N-terminal region (ARP56475_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56475_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Psmc1. It was validated on Western Blot. . Alternate names: AI325227, P26s4, S4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-158