Learn More
Aviva Systems Biology Corp PSMC2 Antibody - N-terminal region (ARP34239_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34239_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PSMC2. Alternate names: MGC3004, MSS1, Nbla10058, S7ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55313