Learn More
Aviva Systems Biology Corp PSMC3IP Antibody - C-terminal region (ARP47615_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47615_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMC3IP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PSMC3IP. Alternate names: GT198, HOP2, TBPIP, ODG3, HUMGT198AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59611