Learn More
Aviva Systems Biology Corp PSMC6 Antibody - C-terminal region (ARP59359_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59359_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMC6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PSMC6. Alternate names: CADP44, MGC12520, P44, SUG2, p42ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-673