Learn More
Aviva Systems Biology Corp PSMD1 Antibody - N-terminal region (ARP56477_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56477_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PSMD1. Alternate names: MGC133040, MGC133041, P112, S1, Rpn2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61877