Learn More
Aviva Systems Biology Corp PSMD11 Antibody - C-terminal region (ARP33002_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33002_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMD11. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human PSMD11. Alternate names: S9, Rpn6, p44.5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-52883