Learn More
Aviva Systems Biology Corp PSMD12 Antibody - N-terminal region (ARP57839_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57839_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMD12. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PSMD12. Alternate names: MGC75406, p55, Rpn5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-400