Learn More
Aviva Systems Biology Corp PSMD14 Antibody - C-terminal region (ARP32449_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32449_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMD14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PSMD14. Alternate names: PAD1, POH1, RPN11ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54596