Learn More
Aviva Systems Biology Corp PSMD3 Antibody - middle region (ARP56480_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56480_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMD3. It was validated on Western Blot. PSMD3,proteasome (prosome, macropain) 26S subunit, non-ATPase, 3,S3; P58; RPN3; TSTA2; PSMD3,26S proteasome non-ATPase regulatory subunit 3,26S proteasome non-ATPase regulatory subunit 3,proteasome subunit p58,26S proteasome regulatory subunit S3,26S proteasome regulatory subunit RPN3,tissue specific transplantation antigen 2The immunogen is a synthetic peptide directed towards the middle region of human PSMD3. Alternate names: P58, RPN3, S3, TSTA2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-160