Learn More
Aviva Systems Biology Corp PSMD4 Antibody - N-terminal region (ARP32342_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32342_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PSMD4. Alternate names: AF, ASF, S5A, AF-1, MCB1, Rpn10, pUB-R5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-54535